Assay Design



Gene Transcripts

Gene Symbol
par-3
Entrez Gene ID
175783
Gene Name
protein-coding
Gene Aliases
CELE_F54E7.3
Chromosome Location
Chr.III: 5664229 - 5676795 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027436.3 NP_001022607.1 5 - 6 509 99
NM_001306739.3 NP_001293668.1 5 - 6 509 99
NM_001306740.3 NP_001293669.1 5 - 6 509 99
NM_001306741.3 NP_001293670.1 5 - 6 509 99
NM_001379719.1 NP_001368259.1 4 - 5 428 99
NM_001379720.2 NP_001367341.1 1 - 2 320 99
NM_001382875.1 5 - 6 505 99
GenBank mRNA U25032.1 - 5 - 6 508 99

View Full Product Details