Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001027436.3 | NP_001022607.1 | 8 - 9 | 2758 | 94 |
NM_001306735.2 | NP_001293664.1 | 4 - 5 | 2440 | 94 | |
NM_001306736.3 | NP_001293665.1 | 4 - 5 | 2440 | 94 | |
NM_001306739.3 | NP_001293668.1 | 8 - 9 | 2758 | 94 | |
NM_001306740.3 | NP_001293669.1 | 8 - 9 | 2758 | 94 | |
NM_001372593.2 | NP_001360234.1 | 4 - 5 | 2440 | 94 | |
NM_001379720.2 | NP_001367341.1 | 4 - 5 | 2569 | 94 | |
NM_001379721.1 | NP_001368258.1 | 4 - 5 | 2440 | 94 | |
NM_001382875.1 | 8 - 9 | 2754 | 94 | ||
GenBank mRNA | U25032.1 | - | 8 - 9 | 2757 | 94 |