Assay Design



Gene Transcripts

Gene Symbol
wrm-1
Entrez Gene ID
175802
Gene Name
protein-coding
Gene Aliases
CELE_B0336.1
Chromosome Location
Chr.III: 5722850 - 5727123 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001312927.3 NP_001299856.1 3 - 4 1410 128
NM_065835.8 NP_498236.1 5 - 6 1605 128
GenBank mRNA AF013951.1 - 5 - 6 1572 128

View Full Product Details