Assay Design



Gene Transcripts

Gene Symbol
eng-1
Entrez Gene ID
184063
Gene Name
protein-coding
Gene Aliases
CELE_F01F1.10
Chromosome Location
Chr.III: 5866495 - 5868501 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026067.5 NP_001021238.1 5 - 6 1083 108
NM_001372601.3 NP_001359901.1 5 - 6 1061 108
NM_065866.6 NP_498267.3 5 - 6 1090 108
GenBank mRNA AB079783.1 - 5 - 6 1083 108

View Full Product Details