Assay Design
Gene Transcripts
- Gene Symbol
- srg-2
- Entrez Gene ID
-
182793
- Gene Name
- Serpentine receptor class gamma-2
- Gene Aliases
-
CELE_C18F10.5
- Chromosome Location
-
Chr.III: 6258590 - 6260215 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065966.2 |
NP_498367.2 |
3 - 4 |
598 |
86 |
View Full Product Details