Assay Design



Gene Transcripts

Gene Symbol
srg-5
Entrez Gene ID
24104887
Gene Name
Serpentine receptor class gamma-5
Gene Aliases
CELE_T12A2.11
Chromosome Location
Chr.III: 6262323 - 6264547 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306705.2 NP_001293634.1 5 - 6 1137 73

View Full Product Details