Assay Design
Gene Transcripts
- Gene Symbol
- T26A5.8
- Entrez Gene ID
-
175921
- Gene Name
- CBFD_NFYB_HMF domain-containing protein
- Gene Aliases
-
CELE_T26A5.8
- Chromosome Location
-
Chr.III: 6462215 - 6463781 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066020.7 |
NP_498421.1 |
1 - 2 |
109 |
81 |
- Gene Symbol
- dlc-1
- Entrez Gene ID
-
175922
- Gene Name
- Dynein light chain 1, cytoplasmic
- Gene Aliases
-
CELE_T26A5.9
- Chromosome Location
-
Chr.III: 6462904 - 6463801 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001382897.1 |
|
1 - 2 |
129 |
81 |
View Full Product Details