Assay Design
Gene Transcripts
- Gene Symbol
- srxa-5
- Entrez Gene ID
-
182448
- Gene Name
- Serpentine Receptor, class XA
- Gene Aliases
-
CELE_C09E7.1
- Chromosome Location
-
Chr.III: 6532947 - 6533819 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066040.3 |
NP_498441.2 |
2 - 3 |
131 |
87 |
View Full Product Details