Assay Design
Gene Transcripts
- Gene Symbol
- isw-1
- Entrez Gene ID
-
175944
- Gene Name
- Chromatin-remodeling complex ATPase chain isw-1
- Gene Aliases
-
CELE_F37A4.8
- Chromosome Location
-
Chr.III: 6703988 - 6710141 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066067.6 |
NP_498468.2 |
10 - 11 |
2485 |
71 |
View Full Product Details