Assay Design



Gene Transcripts

Gene Symbol
kbp-1
Entrez Gene ID
24104812
Gene Name
KNL (kinetochore null) Binding Protein
Gene Aliases
CELE_R13F6.1
Chromosome Location
Chr.III: 6869633 - 6870348 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306710.2 NP_001293639.1 2 - 3 221 100

Gene Symbol
K04C2.8
Entrez Gene ID
3564770
Gene Name
Uncharacterized protein
Gene Aliases
CELE_K04C2.8
Chromosome Location
Chr.III: 6869634 - 6871171 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027489.5 NP_001022660.2 2 - 3 220 100

View Full Product Details