Assay Design
Gene Transcripts
- Gene Symbol
- brc-2
- Entrez Gene ID
-
175962
- Gene Name
- DNA repair protein brc-2
- Gene Aliases
-
CELE_T07E3.5
- Chromosome Location
-
Chr.III: 6900572 - 6903448 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066101.7 |
NP_498502.3 |
11 - 12 |
1291 |
84 |
GenBank mRNA |
AY523518.1 |
- |
10 - 11 |
1173 |
84 |
View Full Product Details