Assay Design
Gene Transcripts
- Gene Symbol
- byn-1
- Entrez Gene ID
-
175968
- Gene Name
- Cell adhesion protein byn-1
- Gene Aliases
-
CELE_F57B9.5
- Chromosome Location
-
Chr.III: 6932001 - 6935868 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066109.5 |
NP_498510.1 |
7 - 8 |
897 |
93 |
View Full Product Details