Assay Design



Gene Transcripts

Gene Symbol
osm-10
Entrez Gene ID
191740
Gene Name
protein-coding
Gene Aliases
CELE_T20H4.1
Chromosome Location
Chr.III: 7243280 - 7245746 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330924.2 NP_001317784.1 6 - 7 1063 121
NM_066197.7 NP_498598.1 5 - 6 1182 121

View Full Product Details