Assay Design
Gene Transcripts
- Gene Symbol
- trd-1
- Entrez Gene ID
-
176055
- Gene Name
- Tetratricopeptide repeat-containing protein trd-1
- Gene Aliases
-
CELE_T20B12.1
- Chromosome Location
-
Chr.III: 7386229 - 7391145 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066235.6 |
NP_498636.2 |
2 - 3 |
145 |
82 |
View Full Product Details