Assay Design



Gene Transcripts

Gene Symbol
lig-4
Entrez Gene ID
176067
Gene Name
LIGase
Gene Aliases
CELE_C07H6.1
Chromosome Location
Chr.III: 7521463 - 7524870 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379799.2 NP_001367462.1 2 - 3 424 79

View Full Product Details