Assay Design
Gene Transcripts
- Gene Symbol
- lin-13
- Entrez Gene ID
-
176083
- Gene Name
- Zinc finger protein lin-13
- Gene Aliases
-
CELE_C03B8.4
- Chromosome Location
-
Chr.III: 7700338 - 7709495 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066277.5 |
NP_498678.3 |
16 - 17 |
5351 |
151 |
GenBank mRNA |
AF245435.1 |
- |
16 - 17 |
5328 |
151 |
View Full Product Details