Assay Design
Gene Transcripts
- Gene Symbol
- tyr-1
- Entrez Gene ID
-
176105
- Gene Name
- Putative tyrosinase-like protein tyr-1
- Gene Aliases
-
CELE_C02C2.1
- Chromosome Location
-
Chr.III: 7870040 - 7877194 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066310.6 |
NP_498711.1 |
8 - 9 |
1424 |
75 |
View Full Product Details