Assay Design
Gene Transcripts
- Gene Symbol
- dcr-1
- Entrez Gene ID
-
176138
- Gene Name
- Death-promoting deoxyribonuclease
- Gene Aliases
-
CELE_K12H4.8
- Chromosome Location
-
Chr.III: 8071967 - 8080281 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066360.4 |
NP_498761.2 |
20 - 21 |
4459 |
106 |
View Full Product Details