Assay Design
Gene Transcripts
- Gene Symbol
- grp-1
- Entrez Gene ID
-
176140
- Gene Name
- GTP exchange factor for ARFs 1
- Gene Aliases
-
CELE_K06H7.4
- Chromosome Location
-
Chr.III: 8085705 - 8088449 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066363.4 |
NP_498764.1 |
5 - 6 |
908 |
105 |
- Gene Symbol
- idi-1
- Entrez Gene ID
-
176142
- Gene Name
- Nudix hydrolase domain-containing protein
- Gene Aliases
-
CELE_K06H7.9
- Chromosome Location
-
Chr.III: 8085699 - 8093046 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001382931.1 |
|
5 - 6 |
921 |
105 |
View Full Product Details