Assay Design
Gene Transcripts
- Gene Symbol
- ncr-2
- Entrez Gene ID
-
176165
- Gene Name
- NPC intracellular cholesterol transporter 1 homolog 2
- Gene Aliases
-
CELE_F09G8.4
- Chromosome Location
-
Chr.III: 8258345 - 8265805 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066412.4 |
NP_498813.1 |
16 - 17 |
3328 |
114 |
View Full Product Details