Assay Design
Gene Transcripts
- Gene Symbol
- rha-2
- Entrez Gene ID
-
176207
- Gene Name
- Putative ATP-dependent RNA helicase rha-2
- Gene Aliases
-
CELE_C06E1.10
- Chromosome Location
-
Chr.III: 8609882 - 8614585 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066494.2 |
NP_498895.1 |
9 - 10 |
2393 |
106 |
View Full Product Details