Assay Design
Gene Transcripts
- Gene Symbol
- twk-7
- Entrez Gene ID
-
192073
- Gene Name
- TWiK family of potassium channels protein 7
- Gene Aliases
-
CELE_F22B7.7
- Chromosome Location
-
Chr.III: 8634067 - 8643739 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066502.6 |
NP_498903.3 |
1 - 2 |
143 |
104 |
View Full Product Details