Assay Design



Gene Transcripts

Gene Symbol
crn-6
Entrez Gene ID
176316
Gene Name
protein-coding
Gene Aliases
CELE_K04H4.6
Chromosome Location
Chr.III: 9367238 - 9370381 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382961.1 6 - 7 712 66
NM_066660.5 NP_499061.2 6 - 7 747 66
GenBank mRNA AY303580.1 - 6 - 7 712 66

View Full Product Details