Assay Design
Gene Transcripts
- Gene Symbol
- crn-6
- Entrez Gene ID
-
176316
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_K04H4.6
- Chromosome Location
-
Chr.III: 9367238 - 9370381 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066660.5 |
NP_499061.2 |
9 - 10 |
1125 |
82 |
View Full Product Details