Assay Design
Gene Transcripts
- Gene Symbol
- emb-30
- Entrez Gene ID
-
176322
- Gene Name
- Abnormal embryogenesis protein 30
- Gene Aliases
-
CELE_F54C8.3
- Chromosome Location
-
Chr.III: 9435507 - 9441539 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066673.7 |
NP_499074.2 |
5 - 6 |
1013 |
105 |
GenBank mRNA |
AF192400.1 |
- |
5 - 6 |
932 |
105 |
View Full Product Details