Assay Design



Gene Transcripts

Gene Symbol
sod-4
Entrez Gene ID
176336
Gene Name
protein-coding
Gene Aliases
CELE_F55H2.1
Chromosome Location
Chr.III: 9496884 - 9500375 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268073.3 NP_001255002.1 4 - 5 410 95
NM_001268074.4 NP_001255003.1 4 - 5 472 95
GenBank mRNA AB003924.1 - 3 - 4 337 95
AB190513.1 - 3 - 4 337 95

View Full Product Details