Assay Design
Gene Transcripts
- Gene Symbol
- srxa-10
- Entrez Gene ID
-
187780
- Gene Name
- Serpentine receptor class XA 10
- Gene Aliases
-
CELE_R10E11.7
- Chromosome Location
-
Chr.III: 9795271 - 9796548 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066770.4 |
NP_499171.3 |
3 - 4 |
408 |
209 |
View Full Product Details