Assay Design
Gene Transcripts
- Gene Symbol
- arl-5
- Entrez Gene ID
-
176392
- Gene Name
- ADP-ribosylation factor-like protein 5
- Gene Aliases
-
CELE_ZK632.8
- Chromosome Location
-
Chr.III: 9819303 - 9824133 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001382971.1 |
|
11 - 12 |
1969 |
86 |
View Full Product Details