Assay Design



Gene Transcripts

Gene Symbol
cyld-1
Entrez Gene ID
176401
Gene Name
protein-coding
Gene Aliases
CELE_F40F12.5
Chromosome Location
Chr.III: 9898575 - 9910658 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268116.2 NP_001255045.1 12 - 13 1491 96
NM_001268117.3 NP_001255046.1 6 - 7 743 96
NM_001268118.3 NP_001255047.1 4 - 5 560 96

View Full Product Details