Assay Design
Gene Transcripts
- Gene Symbol
- ttr-1
- Entrez Gene ID
-
186958
- Gene Name
- Transthyretin-like protein 1
- Gene Aliases
-
CELE_K03H1.6
- Chromosome Location
-
Chr.III: 9929058 - 9930413 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066803.5 |
NP_499204.1 |
1 - 2 |
124 |
65 |
View Full Product Details