Assay Design
Gene Transcripts
- Gene Symbol
- ins-22
- Entrez Gene ID
-
191690
- Gene Name
- Probable insulin-like peptide alpha-type 2
- Gene Aliases
-
CELE_M04D8.2
- Chromosome Location
-
Chr.III: 10026289 - 10026686 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066822.5 |
NP_499223.1 |
1 - 2 |
184 |
83 |
View Full Product Details