Assay Design
Gene Transcripts
- Gene Symbol
- srt-55
- Entrez Gene ID
-
188561
- Gene Name
- Serpentine receptor class T-55
- Gene Aliases
-
CELE_T16H12.8
- Chromosome Location
-
Chr.III: 10104841 - 10106490 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066842.1 |
NP_499243.1 |
1 - 2 |
137 |
74 |
View Full Product Details