Assay Design
Gene Transcripts
- Gene Symbol
- unc-47
- Entrez Gene ID
-
176431
- Gene Name
- Vesicular GABA transporter
- Gene Aliases
-
CELE_T20G5.6
- Chromosome Location
-
Chr.III: 10183757 - 10186260 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066854.6 |
NP_499255.1 |
4 - 5 |
751 |
97 |
GenBank mRNA |
AF031935.1 |
- |
4 - 5 |
751 |
97 |
View Full Product Details