Assay Design
Gene Transcripts
- Gene Symbol
- tbx-8
- Entrez Gene ID
-
176450
- Gene Name
- T-box transcription factor tbx-8
- Gene Aliases
-
CELE_T07C4.2
- Chromosome Location
-
Chr.III: 10347575 - 10349740 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066886.6 |
NP_499287.2 |
1 - 2 |
309 |
96 |
GenBank mRNA |
AB121091.1 |
- |
1 - 2 |
316 |
96 |
View Full Product Details