Assay Design



Gene Transcripts

Gene Symbol
acdh-11
Entrez Gene ID
176477
Gene Name
protein-coding
Gene Aliases
CELE_Y45F3A.3
Chromosome Location
Chr.III: 10566181 - 10571603 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038289.6 NP_001033378.1 6 - 7 1199 102
NM_001379905.1 NP_001367131.1 5 - 6 1084 102

View Full Product Details