Assay Design



Gene Transcripts

Gene Symbol
hpr-9
Entrez Gene ID
176483
Gene Name
protein-coding
Gene Aliases
CELE_Y39A1A.23
Chromosome Location
Chr.III: 10621551 - 10626398 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379907.1 NP_001368495.1 6 - 7 862 105
NM_066941.7 NP_499342.3 6 - 7 940 105

View Full Product Details