Assay Design



Gene Transcripts

Gene Symbol
hpl-2
Entrez Gene ID
176506
Gene Name
protein-coding
Gene Aliases
CELE_K01G5.2
Chromosome Location
Chr.III: 10752195 - 10755530 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027482.4 NP_001022653.1 3 - 4 477 103
NM_001027483.3 NP_001022654.1 3 - 4 468 103
GenBank mRNA AF123573.1 - 3 - 4 462 103

View Full Product Details