Assay Design
Gene Transcripts
- Gene Symbol
- oxi-1
- Entrez Gene ID
-
176515
- Gene Name
- HECT domain-containing protein
- Gene Aliases
-
CELE_Y39A1C.2
- Chromosome Location
-
Chr.III: 10858284 - 10867033 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066991.5 |
NP_499392.1 |
2 - 3 |
310 |
132 |
View Full Product Details