Assay Design



Gene Transcripts

Gene Symbol
strd-1
Entrez Gene ID
176526
Gene Name
STE20-related kinase adapter protein strd-1
Gene Aliases
CELE_Y52D3.1
Chromosome Location
Chr.III: 10947446 - 10951118 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027731.5 NP_001022902.1 4 - 5 891 90
NR_002379.1 4 - 5 865 90

View Full Product Details