Assay Design



Gene Transcripts

Gene Symbol
spat-2
Entrez Gene ID
3564779
Gene Name
protein-coding
Gene Aliases
CELE_Y48A6B.13
Chromosome Location
Chr.III: 11036943 - 11063027 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268172.3 NP_001255101.1 3 - 4 682 55
NM_001347290.2 NP_001334218.1 10 - 11 1731 55
NM_001379924.1 NP_001367286.1 3 - 4 439 55
NM_001379926.1 NP_001366652.1 1 - 2 28 55

View Full Product Details