Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001268172.3 | NP_001255101.1 | 6 - 7 | 1546 | 94 |
NM_001347290.2 | NP_001334218.1 | 13 - 14 | 2595 | 94 | |
NM_001379922.2 | NP_001368618.1 | 14 - 15 | 2637 | 94 | |
NM_001379923.1 | NP_001368619.1 | 6 - 7 | 1553 | 94 | |
NM_001379924.1 | NP_001367286.1 | 6 - 7 | 1303 | 94 | |
NM_001379925.1 | NP_001368620.1 | 13 - 14 | 2577 | 94 | |
NM_001379926.1 | NP_001366652.1 | 4 - 5 | 892 | 94 | |
NM_001379927.1 | NP_001367287.1 | 2 - 3 | 697 | 94 | |
NR_144561.1 | 14 - 15 | 2606 | 94 |