Assay Design
Gene Transcripts
- Gene Symbol
- cku-70
- Entrez Gene ID
-
176546
- Gene Name
- Ku domain-containing protein
- Gene Aliases
-
CELE_Y47D3A.4
- Chromosome Location
-
Chr.III: 11154827 - 11168821 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067037.5 |
NP_499438.2 |
3 - 4 |
514 |
90 |
View Full Product Details