Assay Design
Gene Transcripts
- Gene Symbol
- wht-8
- Entrez Gene ID
-
189976
- Gene Name
- ABC transporter domain-containing protein
- Gene Aliases
-
CELE_Y47D3A.11
- Chromosome Location
-
Chr.III: 11218791 - 11222733 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067041.3 |
NP_499442.2 |
6 - 7 |
650 |
90 |
View Full Product Details