Assay Design



Gene Transcripts

Gene Symbol
obr-1
Entrez Gene ID
176555
Gene Name
protein-coding
Gene Aliases
CELE_Y47D3A.17
Chromosome Location
Chr.III: 11257300 - 11270895 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129258.5 NP_001122730.1 5 - 6 980 88
NM_001379928.1 NP_001367300.1 13 - 14 1829 88
NM_001379929.1 NP_001367172.1 12 - 13 2159 88
NM_001382996.1 13 - 14 1906 88
NM_067047.6 NP_499448.1 13 - 14 1977 88

View Full Product Details