Assay Design



Gene Transcripts

Gene Symbol
obr-1
Entrez Gene ID
176555
Gene Name
protein-coding
Gene Aliases
CELE_Y47D3A.17
Chromosome Location
Chr.III: 11257300 - 11270895 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379928.1 NP_001367300.1 2 - 3 176 62
NM_067047.6 NP_499448.1 2 - 3 324 62

View Full Product Details