Assay Design



Gene Transcripts

Gene Symbol
gly-9
Entrez Gene ID
176557
Gene Name
protein-coding
Gene Aliases
CELE_Y47D3A.23
Chromosome Location
Chr.III: 11278996 - 11290580 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027705.3 NP_001022876.1 7 - 8 1474 76
NM_001382997.1 6 - 7 1106 76

View Full Product Details