Assay Design
Gene Transcripts
- Gene Symbol
- teg-1
- Entrez Gene ID
-
176561
- Gene Name
- Tumorous Enhancer of Glp-1(gf)
- Gene Aliases
-
CELE_Y47D3A.27
- Chromosome Location
-
Chr.III: 11312213 - 11318231 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067054.7 |
NP_499455.2 |
3 - 4 |
598 |
97 |
View Full Product Details