Assay Design
Gene Transcripts
- Gene Symbol
- such-1
- Entrez Gene ID
-
176583
- Gene Name
- Suppressor of spindle checkpoint defect 1
- Gene Aliases
-
CELE_Y66D12A.17
- Chromosome Location
-
Chr.III: 11512850 - 11523759 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067084.6 |
NP_499485.2 |
10 - 11 |
2090 |
115 |
View Full Product Details