Assay Design



Gene Transcripts

Gene Symbol
crh-1
Entrez Gene ID
176597
Gene Name
protein-coding
Gene Aliases
CELE_Y41C4A.4
Chromosome Location
Chr.III: 11657738 - 11695165 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268206.3 NP_001255135.1 1 - 1 422 80

View Full Product Details