Assay Design



Gene Transcripts

Gene Symbol
rsp-8
Entrez Gene ID
176613
Gene Name
protein-coding
Gene Aliases
CELE_C18D11.4
Chromosome Location
Chr.III: 11826418 - 11831006 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268213.3 NP_001255142.1 3 - 4 454 74

View Full Product Details